missing translation for 'onlineSavingsMsg'
Learn More

(2'Z,3'E)-6-Bromoindirubin-3'-oxime 95.0+%, TCI America™

Catalog No. p-7123297
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40065MG 5 mg
B400625MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40065MG Supplier TCI America Supplier No. B40065MG
Only null left

Chemical Identifiers

CAS 667463-62-9
Molecular Formula C16H10BrN3O2
Molecular Weight (g/mol) 356.179
MDL Number MFCD08705318
InChI Key WNWSUJQVZJJGLF-UHFFFAOYSA-N
Synonym bio, 6-bromo-3-3-hydroxyamino indol-2-ylidene-1h-indol-2-one, z,3e-6'-bromo-3-hydroxyimino-1h,1'h-2,3'-biindolyliden-2'-one, 2h-indol-2-one, 6-bromo-3-3e-1,3-dihydro-3-hydroxyimino-2h-indol-2-ylidene-1,3-dihydro-, 3z, 6-bromo-3-3-hydroxyamino indol-2-ylidene indolin-2-one, 2z,3e-6'-bromo-3-hydroxyimino-2,3'-biindolinylidene-2'-one, 6-bromo-3-3-hydroxyamino-2h-indol-2-ylidene-1,3-dihydro-2h-indol-2-one
PubChem CID 24906282
IUPAC Name 6-bromo-3-[3-(hydroxyamino)indol-2-ylidene]-1H-indol-2-one
SMILES C1=CC2=C(C(=C3C4=C(C=C(C=C4)Br)NC3=O)N=C2C=C1)NO

Specifications

Melting Point 300°C
Quantity 5 mg
Formula Weight 356.18
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (2′Z,3′E)-6-Bromoindirubin-3′-oxime
RTECSNumber NM3241900
TSCA No
Recommended Storage Refrigerator