missing translation for 'onlineSavingsMsg'
Learn More

2-Vinylpyridine, 97%, stab. with 0.1% 4-tert-butylcatechol

CAS: 100-69-6 | C7H7N | 105.14 g/mol

Supplier:  Thermo Scientific Chemicals A14056AE

 View more versions of this product

Catalog No. AAA14056AE


Only null left
Explore available promotions

Description

Description

2-Vinylpyridine is a pyridine derivative used in the preparation of poly(2-vinylpyridine) via radical polymerization. 2-Vinylpyridine is used to improve effectiveness of protective polymer coatings on metals. It is a reagent for pyridylethylation for new protection method for active hydrogen compounds.. It is also used as basic materials for one of largest-selling plastic, manufacturing pharmaceuticals and bio-compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Vinylpyridine is a pyridine derivative used in the preparation of poly(2-vinylpyridine) via radical polymerization. 2-Vinylpyridine is used to improve effectiveness of protective polymer coatings on metals. It is a reagent for pyridylethylation for new protection method for active hydrogen compounds.. It is also used as basic materials for one of largest-selling plastic, manufacturing pharmaceuticals and bio-compounds.

Solubility
Soluble in water (27g/L). Slightly soluble in chloroform and methanol.

Notes
Keep container tightly sealed. Store away from oxidizing agent.
Specifications

Specifications

2-Vinylpyridine
100-69-6
0.974
47°C (117°F)
C7H7N
MFCD00006355
UN3073
Light sensitive
Soluble in water (27g/L). Slightly soluble in chloroform and methanol.
C=CC1=CC=CC=N1
105.14
105.14
stabilized with 0.1% 4-tert-butylcatechol
-50°C
159°C
Pungent
1.55
100 mL
104505
2-vinylpyridine, pyridine, 2-ethenyl, pyridine, 2-vinyl, pyridine, ethenyl, vinylpyridine, 2-vinyl pyridine, alpha-vinylpyridine, vinyl pyridine, unii-dt4uv4nnkx, ccris 5238
KGIGUEBEKRSTEW-UHFFFAOYSA-N
2-ethenylpyridine
7521
97%

Chemical Identifiers

100-69-6
105.14
KGIGUEBEKRSTEW-UHFFFAOYSA-N
7521
C=CC1=CC=CC=N1
C7H7N
MFCD00006355
2-vinylpyridine, pyridine, 2-ethenyl, pyridine, 2-vinyl, pyridine, ethenyl, vinylpyridine, 2-vinyl pyridine, alpha-vinylpyridine, vinyl pyridine, unii-dt4uv4nnkx, ccris 5238
2-ethenylpyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H330-H314-H318-H226
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c

H226-H302-H311-H314-H317

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: VINYLPYRIDINES, STABILIZED

EINECSNumber : 202-879-8

RTECSNumber : UU1040000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only