missing translation for 'onlineSavingsMsg'
Learn More

2-Vinylpyridine, 97%, stab. with 0.1% 4-tert-butylcatechol

Catalog No. p-7021644
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA14056AE 100 mL
AAA14056AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA14056AE Supplier Thermo Scientific Chemicals Supplier No. A14056AE
Only null left

CAS: 100-69-6 | C7H7N | 105.14 g/mol

2-Vinylpyridine is a pyridine derivative used in the preparation of poly(2-vinylpyridine) via radical polymerization. 2-Vinylpyridine is used to improve effectiveness of protective polymer coatings on metals. It is a reagent for pyridylethylation for new protection method for active hydrogen compounds.. It is also used as basic materials for one of largest-selling plastic, manufacturing pharmaceuticals and bio-compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Vinylpyridine is a pyridine derivative used in the preparation of poly(2-vinylpyridine) via radical polymerization. 2-Vinylpyridine is used to improve effectiveness of protective polymer coatings on metals. It is a reagent for pyridylethylation for new protection method for active hydrogen compounds.. It is also used as basic materials for one of largest-selling plastic, manufacturing pharmaceuticals and bio-compounds.

Solubility
Soluble in water (27g/L). Slightly soluble in chloroform and methanol.

Notes
Keep container tightly sealed. Store away from oxidizing agent.

Chemical Identifiers

CAS 100-69-6
Molecular Formula C7H7N
Molecular Weight (g/mol) 105.14
MDL Number MFCD00006355
InChI Key KGIGUEBEKRSTEW-UHFFFAOYSA-N
Synonym 2-vinylpyridine, pyridine, 2-ethenyl, pyridine, 2-vinyl, pyridine, ethenyl, vinylpyridine, 2-vinyl pyridine, alpha-vinylpyridine, vinyl pyridine, unii-dt4uv4nnkx, ccris 5238
PubChem CID 7521
IUPAC Name 2-ethenylpyridine
SMILES C=CC1=CC=CC=N1

Specifications

Melting Point -50°C
Density 0.974
Boiling Point 159°C
Flash Point 47°C (117°F)
Odor Pungent
Refractive Index 1.55
Quantity 100 mL
UN Number UN3073
Beilstein 104505
Sensitivity Light sensitive
Solubility Information Soluble in water (27g/L). Slightly soluble in chloroform and methanol.
Formula Weight 105.14
Percent Purity 97%
Chemical Name or Material 2-Vinylpyridine, stabilized with 0.1% 4-tert-butylcatechol
Show More Show Less
Hazard Category H226-H302-H311-H314-H317
Hazard Statement GHS H Statement
H301-H311-H330-H314-H318-H226
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: VINYLPYRIDINES, STABILIZED
EINECSNumber 202-879-8
RTECSNumber UU1040000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only