missing translation for 'onlineSavingsMsg'
Learn More

2-Vinylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. V01401G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V01401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V01401G Supplier TCI America Supplier No. V01401G

Specifications

Chemical Name or Material 2-Vinylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 15016-42-9
Melting Point 109°C
Color White-Yellow
Molecular Formula C8H9BO2
Quantity 1 g
MDL Number MFCD01074659
Synonym 2-vinylphenylboronic acid, 2-vinylbenzeneboronic acid, 2-vinylphenyl boronic acid, 2-ethenylphenyl boronic acid, 2-boronostyrene, styrene-2-boronic acid, boronic acid, ethenylphenyl, 2-vinylphenylboronicacid, acmc-1bt6g, 2-vinyl-phenylboronic acid
InChI Key QHFAXRHEKNHTDH-UHFFFAOYSA-N
SMILES B(C1=CC=CC=C1C=C)(O)O
IUPAC Name (2-ethenylphenyl)boronic acid
Molecular Weight (g/mol) 147.968
PubChem CID 2773587
Formula Weight 147.97
Physical Form Crystalline Powder
Show More Show Less