missing translation for 'onlineSavingsMsg'
Learn More

2-Vinyloxytetrahydropyran 97.0+%, TCI America™

Catalog No. v00765g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V00765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V00765G Supplier TCI America Supplier No. V00765G
Only null left

Chemical Identifiers

CAS 22408-41-9
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD03093644
InChI Key BYUNYALHUMSCSA-UHFFFAOYSA-N
Synonym Tetrahydro-2-vinyloxy-2H-pyran
PubChem CID 11528345
IUPAC Name 2-ethenoxyoxane
SMILES C=COC1CCCCO1

Specifications

Color Yellow
Boiling Point 60°C
Quantity 5 g
UN Number 1993
Formula Weight 128.17
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Vinyloxytetrahydropyran
TSCA No
Recommended Storage Refrigerator