missing translation for 'onlineSavingsMsg'
Learn More

2-Vinylnaphthalene, 97%

CAS: 827-54-3 | C12H10 | 154.21 g/mol

Supplier:  Thermo Scientific Chemicals A1247014

 View more versions of this product

Catalog No. AAA1247014

Only null left
Explore available promotions

Description

Description

2-Vinylnaphthalene is employed in the synthesis of various copolymers such as Poly (sodium styrenesulfonate-co-2-vinyl-napthalene) copolymer and Poly (2-vinylnapthalene-alt-maleic acid)-graft-polystyrene. The latter finds applications as a photoactive polymer micelle and stabilizer for polystyrene latexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Vinylnaphthalene is employed in the synthesis of various copolymers such as Poly (sodium styrenesulfonate-co-2-vinyl-napthalene) copolymer and Poly (2-vinylnapthalene-alt-maleic acid)-graft-polystyrene. The latter finds applications as a photoactive polymer micelle and stabilizer for polystyrene latexes.

Solubility
Soluble in ethanol, acetone, and benzene. Insoluble in water.

Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents. Protect away from heat.
Specifications

Specifications

2-Vinylnaphthalene
62°C to 67°C
>110°C (230°F)
MFCD00004125
2038190
Soluble in ethanol,acetone,and benzene. Insoluble in water.
C=CC1=CC=C2C=CC=CC2=C1
154.21
CHEBI:51325
97%
827-54-3
128°C to 131°C (13 mmHg)
C12H10
25 g
2-vinylnaphthalene, naphthalene, 2-ethenyl, 2-vinyl-naphthalene, beta-vinylnaphthalene, poly 2-vinylnaphthalene, unii-hzd8li91n1, hzd8li91n1, polyvinylnaphthalene, 2-vinyl naphthalene, 2-ethenyinaphthalene
KXYAVSFOJVUIHT-UHFFFAOYSA-N
2-ethenylnaphthalene
13230
154.21

Chemical Identifiers

827-54-3
154.21
KXYAVSFOJVUIHT-UHFFFAOYSA-N
13230
2-ethenylnaphthalene
C12H10
MFCD00004125
2-vinylnaphthalene, naphthalene, 2-ethenyl, 2-vinyl-naphthalene, beta-vinylnaphthalene, poly 2-vinylnaphthalene, unii-hzd8li91n1, hzd8li91n1, polyvinylnaphthalene, 2-vinyl naphthalene, 2-ethenyinaphthalene
CHEBI:51325
C=CC1=CC=C2C=CC=CC2=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H315-H319-H317-H335-H208
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 212-573-6

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only