missing translation for 'onlineSavingsMsg'
Learn More

2-Vinylnaphthalene, 97%

Catalog No. p-7020491
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1247006 5 g
AAA1247014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1247006 Supplier Thermo Scientific Chemicals Supplier No. A1247006
Only null left

CAS: 827-54-3 | C12H10 | 154.21 g/mol

2-Vinylnaphthalene is employed in the synthesis of various copolymers such as Poly (sodium styrenesulfonate-co-2-vinyl-napthalene) copolymer and Poly (2-vinylnapthalene-alt-maleic acid)-graft-polystyrene. The latter finds applications as a photoactive polymer micelle and stabilizer for polystyrene latexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Vinylnaphthalene is employed in the synthesis of various copolymers such as Poly (sodium styrenesulfonate-co-2-vinyl-napthalene) copolymer and Poly (2-vinylnapthalene-alt-maleic acid)-graft-polystyrene. The latter finds applications as a photoactive polymer micelle and stabilizer for polystyrene latexes.

Solubility
Soluble in ethanol, acetone, and benzene. Insoluble in water.

Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents. Protect away from heat.

Chemical Identifiers

CAS 827-54-3
Molecular Formula C12H10
Molecular Weight (g/mol) 154.21
MDL Number MFCD00004125
InChI Key KXYAVSFOJVUIHT-UHFFFAOYSA-N
Synonym 2-vinylnaphthalene, naphthalene, 2-ethenyl, 2-vinyl-naphthalene, beta-vinylnaphthalene, poly 2-vinylnaphthalene, unii-hzd8li91n1, hzd8li91n1, polyvinylnaphthalene, 2-vinyl naphthalene, 2-ethenyinaphthalene
PubChem CID 13230
ChEBI CHEBI:51325
IUPAC Name 2-ethenylnaphthalene
SMILES C=CC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 62°C to 67°C
Boiling Point 128°C to 131°C (13 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2038190
Solubility Information Soluble in ethanol,acetone,and benzene. Insoluble in water.
Formula Weight 154.21
Percent Purity 97%
Chemical Name or Material 2-Vinylnaphthalene
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H332-H315-H319-H317-H335-H208
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 212-573-6
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only