missing translation for 'onlineSavingsMsg'
Learn More

2-Undecylimidazole 96.0+%, TCI America™

Catalog No. p-7137840
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
U001225G 25 g
U0012500G 500 g
2 options

Catalog No. U001225G

Supplier: TCI America U001225G

Only null left

Chemical Identifiers

CAS 16731-68-3
Molecular Formula C14H26N2
Molecular Weight (g/mol) 222.376
MDL Number MFCD00038694
InChI Key LLEASVZEQBICSN-UHFFFAOYSA-N
PubChem CID 85582
IUPAC Name 2-undecyl-1H-imidazole
SMILES CCCCCCCCCCCC1=NC=CN1

Specifications

Melting Point 73°C
Color White
Boiling Point 215°C
Quantity 25 g
Formula Weight 222.38
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Undecylimidazole
EINECSNumber (5)-0383
TSCA Yes