missing translation for 'onlineSavingsMsg'
Learn More

2-(Triphenylphosphoranylidene)acetophenone 98.0+%, TCI America™

Catalog No. p-7137059
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20021G 1 g
T20025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20021G Supplier TCI America Supplier No. T20021G
Only null left

Chemical Identifiers

CAS 859-65-4
Molecular Formula C26H21OP
Molecular Weight (g/mol) 380.43
MDL Number MFCD00014088
InChI Key MZRSAJZDYIISJW-UHFFFAOYSA-N
Synonym benzoylmethylene triphenylphosphorane, 2-triphenylphosphoranylidene acetophenone, 1-phenyl-2-triphenylphosphoranylidene ethanone, phenacylidenetriphenyl phosphorane, phenylcarbonylmethylene triphenylphosphorane, acetophenone, 2-triphenylphosphoranylidene, phenacylidenetriphenylphosphorane, triphenylphosphinebenzoylmethylene, ethanone, 1-phenyl-2-triphenylphosphoranylidene
PubChem CID 70073
IUPAC Name 1-phenyl-2-(triphenyl-λ⁵-phosphanylidene)ethan-1-one
SMILES O=C(C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 1 g
Formula Weight 380.43
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Triphenylphosphoranylidene)acetophenone
RTECSNumber AN0540000
TSCA No
Recommended Storage Refrigerator