missing translation for 'onlineSavingsMsg'
Learn More

2-Triphenylenecarboxaldehyde 98.0+%, TCI America™

Catalog No. T3304200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3304200MG 200 mg
T33041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3304200MG Supplier TCI America Supplier No. T3304200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 96404-79-4
Molecular Formula C19H12O
Molecular Weight (g/mol) 256.30
MDL Number MFCD29089374
InChI Key NXDGVEYXRSKONY-UHFFFAOYSA-N
PubChem CID 13375435
IUPAC Name triphenylene-2-carbaldehyde
SMILES O=CC1=CC=C2C3=CC=CC=C3C3=CC=CC=C3C2=C1

Specifications

Melting Point 162°C
Color White
Quantity 200 mg
Formula Weight 256.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Triphenylenecarboxaldehyde