missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Trimethylsilylthiazole 95.0+%, TCI America™
$115.57 - $456.70
Chemical Identifiers
| CAS | 79265-30-8 |
|---|---|
| Molecular Formula | C6H11NSSi |
| Molecular Weight (g/mol) | 157.31 |
| MDL Number | MFCD00066274 |
| InChI Key | VJCHUDDPWPQOLH-UHFFFAOYSA-N |
| Synonym | 2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq |
| PubChem CID | 588453 |
| IUPAC Name | 2-(trimethylsilyl)-1,3-thiazole |
| SMILES | C[Si](C)(C)C1=NC=CS1 |
Chemical Identifiers
| 79265-30-8 | |
| 157.31 | |
| VJCHUDDPWPQOLH-UHFFFAOYSA-N | |
| 588453 | |
| C[Si](C)(C)C1=NC=CS1 |
| C6H11NSSi | |
| MFCD00066274 | |
| 2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq | |
| 2-(trimethylsilyl)-1,3-thiazole |
Specifications
| 79265-30-8 | |
| 57°C | |
| MFCD00066274 | |
| 2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq | |
| C[Si](C)(C)C1=NC=CS1 | |
| 157.31 | |
| 157.31 | |
| Liquid |
| Yellow | |
| C6H11NSSi | |
| 1 mL | |
| VJCHUDDPWPQOLH-UHFFFAOYSA-N | |
| 2-(trimethylsilyl)-1,3-thiazole | |
| 588453 | |
| ≥95.0% (GC) | |
| 2-Trimethylsilylthiazole |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator