missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trimethylsilyloxy)furan 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 50°C |
| Quantity | 5 g |
| UN Number | 1993 |
| Formula Weight | 156.26 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-(Trimethylsilyloxy)furan |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |