CAS RN 61550-02-5
IUPAC Name:
(furan-2-yloxy)trimethylsilane
Synonyms:
(furan-2-yloxy)trimethylsilane
Molecular Weight (g/mol):
156.26
Molecular Formula:
C7H12O2Si
InChi Key:
ILBCDLOQMRDXLN-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)OC1=CC=CO1
1
–
1
of
1
results
1
2-(Trimethylsilyloxy)furan 96.0+%, TCI America™
CAS: 61550-02-5 Molecular Formula: C7H12O2Si Molecular Weight (g/mol): 156.26 MDL Number: MFCD00011729 InChI Key: ILBCDLOQMRDXLN-UHFFFAOYSA-N Synonym: (2-Furanyloxy)trimethylsilane PubChem CID: 188414 IUPAC Name: (furan-2-yloxy)trimethylsilane SMILES: C[Si](C)(C)OC1=CC=CO1