missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyloxy)-1,3-cyclohexadiene 94.0+%, TCI America™

Catalog No. T26305G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T26305G Supplier TCI America Supplier No. T26305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54781-19-0
Molecular Formula C9H16OSi
Molecular Weight (g/mol) 168.311
MDL Number MFCD00274202
InChI Key WPIRVUXAMPRMAY-UHFFFAOYSA-N
Synonym 1-(Trimethylsilyloxy)-3,4-dihydrobenzene
PubChem CID 4095656
IUPAC Name cyclohexa-1,5-dien-1-yloxy(trimethyl)silane
SMILES C[Si](C)(C)OC1=CCCC=C1

Specifications

Color Yellow
Boiling Point 60°C
Quantity 5 g
UN Number 1993
Formula Weight 168.31
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trimethylsilyloxy)-1,3-cyclohexadiene
TSCA No
Recommended Storage Refrigerator