missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trimethylsilylethynyl)thiophene 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 90°C |
| Quantity | 5 g |
| UN Number | 1993 |
| Formula Weight | 180.34 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-(Trimethylsilylethynyl)thiophene |
Safety and Handling
| TSCA | No |