accessibility menu, dialog, popup

UserName

CH3SiH3CCH3S

CAS RN 40231-03-6

IUPAC Name: trimethyl[2-(thiophen-2-yl)ethynyl]silane
Synonyms: trimethyl[2-(thiophen-2-yl)ethynyl]silane
Molecular Weight (g/mol): 180.34
Molecular Formula: C9H12SSi
InChi Key: OQUBLKNISPLGJP-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CC=CS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results