missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)thiophene, 97%

CAS: 18245-28-8 | C7H12SSi | 156.318 g/mol

Supplier:  Thermo Scientific Chemicals H6117014

 View more versions of this product

Catalog No. AAH6117014


Only null left
Explore available promotions

Description

Description

2-(Trimethylsilyl)thiophene is used to produce 2,5-bis-adamantan-1-yl-thiophene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trimethylsilyl)thiophene is used to produce 2,5-bis-adamantan-1-yl-thiophene.

Notes
Keep container tightly sealed. Store refrigerated conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-(Trimethylsilyl)thiophene
0.926
C7H12SSi
MFCD00005415
UN1993
OANGLGSZPSFVDY-UHFFFAOYSA-N
trimethyl(thiophen-2-yl)silane
140360
97%
18245-28-8
41°C (106°F)
1.497
25 g
2-thienyltrimethylsilane, 2-trimethylsilylthiophene, trimethyl thiophen-2-yl silane, silane, trimethyl-2-thienyl, trimethyl-2-thienylsilane, 2-trimethylsilyl thiophene, trimethyl 2-thienyl silane, amtsi021, acmc-1c71q, thien-2-yl-trimethyl-silane
C[Si](C)(C)C1=CC=CS1
156.318
156.32

Chemical Identifiers

18245-28-8
156.318
OANGLGSZPSFVDY-UHFFFAOYSA-N
140360
C[Si](C)(C)C1=CC=CS1
C7H12SSi
MFCD00005415
2-thienyltrimethylsilane, 2-trimethylsilylthiophene, trimethyl thiophen-2-yl silane, silane, trimethyl-2-thienyl, trimethyl-2-thienylsilane, 2-trimethylsilyl thiophene, trimethyl 2-thienyl silane, amtsi021, acmc-1c71q, thien-2-yl-trimethyl-silane
trimethyl(thiophen-2-yl)silane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only