missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)thiophene, 97%

Catalog No. p-7041162
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6117006 5 g
AAH6117014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6117006 Supplier Thermo Scientific Chemicals Supplier No. H6117006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 18245-28-8 | C7H12SSi | 156.318 g/mol

2-(Trimethylsilyl)thiophene is used to produce 2,5-bis-adamantan-1-yl-thiophene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trimethylsilyl)thiophene is used to produce 2,5-bis-adamantan-1-yl-thiophene.

Notes
Keep container tightly sealed. Store refrigerated conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 18245-28-8
Molecular Formula C7H12SSi
Molecular Weight (g/mol) 156.318
MDL Number MFCD00005415
InChI Key OANGLGSZPSFVDY-UHFFFAOYSA-N
Synonym 2-thienyltrimethylsilane, 2-trimethylsilylthiophene, trimethyl thiophen-2-yl silane, silane, trimethyl-2-thienyl, trimethyl-2-thienylsilane, 2-trimethylsilyl thiophene, trimethyl 2-thienyl silane, amtsi021, acmc-1c71q, thien-2-yl-trimethyl-silane
PubChem CID 140360
IUPAC Name trimethyl(thiophen-2-yl)silane
SMILES C[Si](C)(C)C1=CC=CS1

Specifications

Density 0.926
Flash Point 41°C (106°F)
Refractive Index 1.497
Quantity 5 g
UN Number UN1993
Formula Weight 156.32
Percent Purity 97%
Chemical Name or Material 2-(Trimethylsilyl)thiophene
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only