Learn More
2-(Trimethylsilyl)thiazole, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 292290250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 2-(Trimethylsilyl)thiazole | |
| 95.0 | |
| 0.9800g/mL | |
| 62°C | |
| 95% min. (GC) | |
| C6H11NSSi | |
| MFCD00066274 | |
| 0.98 | |
| Solubility in water: insoluble | |
| C[Si](C)(C)C1=NC=CS1 | |
| 157.31 | |
| 157.31 |
| 79265-30-8 | |
| 100.0 | |
| 53.0°C to 55.0°C (10.0mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4950 to 1.5000 | |
| 25mL | |
| 2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq | |
| VJCHUDDPWPQOLH-UHFFFAOYSA-N | |
| trimethyl(1,3-thiazol-2-yl)silane | |
| 588453 | |
| 96% |
Chemical Identifiers
| 79265-30-8 | |
| 157.31 | |
| VJCHUDDPWPQOLH-UHFFFAOYSA-N | |
| 588453 | |
| C[Si](C)(C)C1=NC=CS1 |
| C6H11NSSi | |
| MFCD00066274 | |
| 2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq | |
| trimethyl(1,3-thiazol-2-yl)silane |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF exposed or concerned: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO â Research Use Only