missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)thiazole, 96%

Catalog No. AC292290050
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC292290050 5 mL Glass bottle
AC292290250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC292290050 Supplier Thermo Scientific Chemicals Supplier No. 292290050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 79265-30-8 | C6H11NSSi | 157.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79265-30-8
Molecular Formula C6H11NSSi
Molecular Weight (g/mol) 157.31
MDL Number MFCD00066274
InChI Key VJCHUDDPWPQOLH-UHFFFAOYSA-N
Synonym 2-trimethylsilyl thiazole, 2-trimethylsilylthiazole, 2-trimethylsilyl-1,3-thiazole, thiazole, 2-trimethylsilyl, 2-trimethylsilanylthiazole, trimethyl 1,3-thiazol-2-yl silane, 2-tms-thiazole, pubchem10262, acmc-209pgq
PubChem CID 588453
IUPAC Name trimethyl(1,3-thiazol-2-yl)silane
SMILES C[Si](C)(C)C1=NC=CS1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9800g/mL
Boiling Point 53.0°C to 55.0°C (10.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4950 to 1.5000
Quantity 5 mL
Specific Gravity 0.98
Solubility Information Solubility in water: insoluble
Formula Weight 157.31
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-(Trimethylsilyl)thiazole
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only