missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)phenyl Trifluoromethanesulfonate 95.0+%, TCI America™

Supplier:  TCI America T208925G

 View more versions of this product

Catalog No. T208925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-(Trimethylsilyl)phenyl Trifluoromethanesulfonate
Yellow
C10H13F3O3SSi
25 g
2-(Trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 2-(Trimethylsilyl)phenyl Ester
C[Si](C)(C)C1=CC=CC=C1OS(=O)(=O)C(F)(F)F
298.351
298.35
Liquid
88284-48-4
70°C
MFCD00799598
3265
XBHPFCIWRHJDCP-UHFFFAOYSA-N
(2-trimethylsilylphenyl) trifluoromethanesulfonate
3384007
≥95.0% (GC)

Chemical Identifiers

88284-48-4
298.351
XBHPFCIWRHJDCP-UHFFFAOYSA-N
3384007
C[Si](C)(C)C1=CC=CC=C1OS(=O)(=O)C(F)(F)F
C10H13F3O3SSi
MFCD00799598
2-(Trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 2-(Trimethylsilyl)phenyl Ester
(2-trimethylsilylphenyl) trifluoromethanesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions