missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)phenyl Trifluoromethanesulfonate 95.0+%, TCI America™

Catalog No. T20895G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20895G 5 g
T20891G 1 g
T208925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T20895G Supplier TCI America Supplier No. T20895G
Only null left

Chemical Identifiers

CAS 88284-48-4
Molecular Formula C10H13F3O3SSi
Molecular Weight (g/mol) 298.351
MDL Number MFCD00799598
InChI Key XBHPFCIWRHJDCP-UHFFFAOYSA-N
Synonym 2-(Trimethylsilyl)phenyl Triflate, Trifluoromethanesulfonic Acid 2-(Trimethylsilyl)phenyl Ester
PubChem CID 3384007
IUPAC Name (2-trimethylsilylphenyl) trifluoromethanesulfonate
SMILES C[Si](C)(C)C1=CC=CC=C1OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 70°C
Quantity 5 g
UN Number 3265
Formula Weight 298.35
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trimethylsilyl)phenyl Trifluoromethanesulfonate