missing translation for 'onlineSavingsMsg'
Learn More

2-[(Trimethylsilyl)ethynyl]aniline, 97%

Catalog No. p-7052810
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5593003 1 g
AAH5593006 5 g
AAH5593014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5593003 Supplier Thermo Scientific Chemicals Supplier No. H5593003

CAS: 103529-16-4 | C11H15NSi | 189.333 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103529-16-4
Molecular Formula C11H15NSi
Molecular Weight (g/mol) 189.333
MDL Number MFCD02093646
InChI Key ISXSBRZDPKQYSC-UHFFFAOYSA-N
Synonym 2-trimethylsilyl ethynyl aniline, 2-2-trimethylsilylethynyl aniline, 2-trimethylsilylethynyl aniline, 2-2-trimethylsilyl ethynyl aniline, benzenamine, 2-trimethylsilyl ethynyl
PubChem CID 4377953
IUPAC Name 2-(2-trimethylsilylethynyl)aniline
SMILES C[Si](C)(C)C#CC1=CC=CC=C1N

Specifications

Color Colorless to Yellow
Density 0.94 g/mL
Boiling Point 120°C (0.1 mmHg)
Flash Point 110°C (230°F)
Refractive Index 1.564
Quantity 1 g
Formula Weight 189.33
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-[(Trimethylsilyl)ethynyl]aniline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only