accessibility menu, dialog, popup

UserName

CAS RN 103529-16-4

IUPAC Name: 2-[2-(trimethylsilyl)ethynyl]aniline
Synonyms: 2-[2-(trimethylsilyl)ethynyl]aniline
Molecular Weight (g/mol): 189.33
Molecular Formula: C11H15NSi
InChi Key: ISXSBRZDPKQYSC-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CC1=CC=CC=C1N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)