missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)ethoxymethyl chloride, 90%, tech., stabilized

Catalog No. AC219020050
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC219020050 5 mL Glass bottle
AC219020250 25 mL Glass bottle
AC219021000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC219020050 Supplier Thermo Scientific Chemicals Supplier No. 219020050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 76513-69-4 | C6H15ClOSi | 166.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76513-69-4
Molecular Formula C6H15ClOSi
Molecular Weight (g/mol) 166.72
InChI Key BPXKZEMBEZGUAH-UHFFFAOYSA-N
Synonym 2-trimethylsilyl ethoxymethyl chloride, 2-chloromethoxy ethyl trimethylsilane, sem-chloride, 2-trimethylsilyl ethoxymethylchloride, sem-cl, 2-chloromethyl 2-trimethylsilyl ethyl ether, semcl, 2-chloromethoxy ethyl-trimethylsilane, 2-trimethysilyl-ethoxymethyl chloride
PubChem CID 2724271
IUPAC Name 2-(chloromethoxy)ethyl-trimethylsilane
SMILES C[Si](C)(C)CCOCCl

Specifications

Color Colorless
Density 0.9400g/mL
Boiling Point 57.0°C to 59.0°C (8.0 mmHg)
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.4350 to 1.4370
Linear Formula ClCH2O(CH2)2Si(CH3)3
Quantity 5 mL
Fieser 10,431; 16,361; 01,107
Specific Gravity 0.94
Solubility Information Solubility in water: decomposes
Stabilizer 10ppm diisopropylethylamine
Formula Weight 166.72
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 2-(Trimethylsilyl)ethoxymethyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 278-483-4

RUO – Research Use Only