accessibility menu, dialog, popup

UserName

CH3SiCH3CH3OCl

CAS RN 76513-69-4

IUPAC Name: [2-(chloromethoxy)ethyl]trimethylsilane
Synonyms: [2-(chloromethoxy)ethyl]trimethylsilane
Molecular Weight (g/mol): 166.72
Molecular Formula: C6H15ClOSi
InChi Key: BPXKZEMBEZGUAH-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CCOCCl
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)