Learn More
2-(Trimethylsilyl)ethanol, 98+%
CAS: 2916-68-9 | C5H14OSi | 118.251 g/mol
Supplier: Thermo Scientific Chemicals B2097022
Description
2-(Trimethylsilyl)ethanol is used as a protecting reagent for carboxyl, phosphoryl, hydroxyl and amino group in organic synthesis. It is used as a precursor to prepare trimethyl(2-phenoxyethyl)silanes by reacting with aromatic fluoride. It is also used in the synthesis of teoc-protected amines by using the corresponding isocyanates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(Trimethylsilyl)ethanol is used as a protecting reagent for carboxyl, phosphoryl, hydroxyl and amino group in organic synthesis. It is used as a precursor to prepare trimethyl(2-phenoxyethyl)silanes by reacting with aromatic fluoride. It is also used in the synthesis of teoc-protected amines by using the corresponding isocyanates.
Solubility
Miscible with water.
Notes
Keep away from sources of ignition. Keep container tightly closed in a dry and well-ventilated place.
Specifications
| 2-(Trimethylsilyl)ethanol | |
| 0.825 | |
| 50°C (122°F) | |
| C5H14OSi | |
| MFCD00002825 | |
| UN1987 | |
| 2-trimethylsilyl ethanol, ethanol, 2-trimethylsilyl, 2-hydroxyethyl trimethylsilane, 2-trimethylsilyl ethan-1-ol, silane, 2-hydroxyethyl trimethyl, 2-trimethylsilanyl-ethanol, 2-trimethylsilyl-ethanol, 2-trimethylsilyl ethanal | |
| ZNGINKJHQQQORD-UHFFFAOYSA-N | |
| 2-trimethylsilylethanol | |
| 18013 | |
| ≥98% |
| 2916-68-9 | |
| 71°C to 72°C (10 mmHg) | |
| 98+% | |
| 1.425 | |
| 100 g | |
| 1732034 | |
| Miscible with water. | |
| C[Si](C)(C)CCO | |
| 118.251 | |
| 118.25 |
Chemical Identifiers
| 2916-68-9 | |
| 118.251 | |
| ZNGINKJHQQQORD-UHFFFAOYSA-N | |
| 18013 | |
| C[Si](C)(C)CCO |
| C5H14OSi | |
| MFCD00002825 | |
| 2-trimethylsilyl ethanol, ethanol, 2-trimethylsilyl, 2-hydroxyethyl trimethylsilane, 2-trimethylsilyl ethan-1-ol, silane, 2-hydroxyethyl trimethyl, 2-trimethylsilanyl-ethanol, 2-trimethylsilyl-ethanol, 2-trimethylsilyl ethanal | |
| 2-trimethylsilylethanol |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 220-844-5
RTECSNumber : KM5480000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only