accessibility menu, dialog, popup

UserName

CAS RN 2916-68-9

IUPAC Name: 2-(trimethylsilyl)ethan-1-ol
Synonyms: 2-(trimethylsilyl)ethanol
Molecular Weight (g/mol): 118.25
Molecular Formula: C5H14OSi
InChi Key: ZNGINKJHQQQORD-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CCO
Boiling Point 
  • (1)
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (1)