missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Trimethylsilyl-1,3-dithiane 96.0+%, TCI America™
Supplier: TCI America T151425G
Specifications
| 2-Trimethylsilyl-1,3-dithiane | |
| Yellow | |
| C7H16S2Si | |
| 25 g | |
| BTTUMVHWIAXYPJ-UHFFFAOYSA-N | |
| (1,3-dithian-2-yl)trimethylsilane | |
| 83413 | |
| ≥96.0% (GC) |
| 13411-42-2 | |
| 77°C | |
| MFCD00006655 | |
| 2-trimethylsilyl-1,3-dithiane, 1,3-dithian-2-yl trimethylsilane, 1,3-dithian-2-yl trimethyl silane, silane, 1,3-dithian-2-yltrimethyl, zlchem 985, acmc-1bxzr, 2-trimethylsily-1,3-dithiane, 2-trimethylsilyl-1, 3-dithiane, 2-trimethysilyl-1,3-dithiane | |
| C[Si](C)(C)C1SCCCS1 | |
| 192.41 | |
| 192.41 | |
| Liquid |
Chemical Identifiers
| 13411-42-2 | |
| 192.41 | |
| BTTUMVHWIAXYPJ-UHFFFAOYSA-N | |
| 83413 | |
| C[Si](C)(C)C1SCCCS1 |
| C7H16S2Si | |
| MFCD00006655 | |
| 2-trimethylsilyl-1,3-dithiane, 1,3-dithian-2-yl trimethylsilane, 1,3-dithian-2-yl trimethyl silane, silane, 1,3-dithian-2-yltrimethyl, zlchem 985, acmc-1bxzr, 2-trimethylsily-1,3-dithiane, 2-trimethylsilyl-1, 3-dithiane, 2-trimethysilyl-1,3-dithiane | |
| (1,3-dithian-2-yl)trimethylsilane |
Safety and Handling
TSCA : No