accessibility menu, dialog, popup

UserName

CH3SiH3CCH3SS

CAS RN 13411-42-2

IUPAC Name: (1,3-dithian-2-yl)trimethylsilane
Synonyms: 1,3-dithian-2-yltrimethylsilane
Molecular Weight (g/mol): 192.41
Molecular Formula: C7H16S2Si
InChi Key: BTTUMVHWIAXYPJ-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1SCCCS1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results