Learn More
2-(Trimethylsiloxy)ethyl methacrylate, 94%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
$224.19 - $224.19
Chemical Identifiers
| CAS | 17407-09-9 |
|---|---|
| Molecular Formula | C9H18O3Si |
| Molecular Weight (g/mol) | 202.33 |
| MDL Number | MFCD00053869 |
| InChI Key | WUGOQZFPNUYUOO-UHFFFAOYSA-N |
| Synonym | 2-trimethylsilyl oxy ethyl methacrylate, 2-trimethylsilyloxy ethyl methacrylate, methacryloxyethoxytrimethylsilane, 2-trimethylsiloxy ethyl methacrylate, trimethylsiloxy-ethylmethacrylate, acmc-1buxw, trimethylsilyloxyethyl-methacrylate, 2-trimethylsiloxyethyl methacrylate, 2-trimethylsiloxy ethylmethacrylate |
| PubChem CID | 87094 |
| IUPAC Name | 2-trimethylsilyloxyethyl 2-methylprop-2-enoate |
| SMILES | CC(=C)C(=O)OCCO[Si](C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1667518
|
Thermo Scientific Chemicals
L1667518 |
50 g |
Each for $224.19
|
|
|||||
Description
2-(Trimethylsiloxy)ethyl methacrylate is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(Trimethylsiloxy)ethyl methacrylate is used as pharmaceutical intermediates.
Solubility
Difficult to mix.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 17407-09-9 | |
| 202.33 | |
| WUGOQZFPNUYUOO-UHFFFAOYSA-N | |
| 87094 | |
| CC(=C)C(=O)OCCO[Si](C)(C)C |
| C9H18O3Si | |
| MFCD00053869 | |
| 2-trimethylsilyl oxy ethyl methacrylate, 2-trimethylsilyloxy ethyl methacrylate, methacryloxyethoxytrimethylsilane, 2-trimethylsiloxy ethyl methacrylate, trimethylsiloxy-ethylmethacrylate, acmc-1buxw, trimethylsilyloxyethyl-methacrylate, 2-trimethylsiloxyethyl methacrylate, 2-trimethylsiloxy ethylmethacrylate | |
| 2-trimethylsilyloxyethyl 2-methylprop-2-enoate |
Specifications
| 17407-09-9 | |
| 65°C (0.9 mmHg) | |
| C9H18O3Si | |
| MFCD00053869 | |
| Moisture sensitive | |
| Difficult to mix. | |
| CC(=C)C(=O)OCCO[Si](C)(C)C | |
| 202.33 | |
| 202.33 | |
| 2-(Trimethylsiloxy)ethyl methacrylate, stabilized with 100ppm 4-methoxyphenol |
| 0.995 | |
| 42°C (107°F) | |
| 1.43 | |
| 50 g | |
| 2-trimethylsilyl oxy ethyl methacrylate, 2-trimethylsilyloxy ethyl methacrylate, methacryloxyethoxytrimethylsilane, 2-trimethylsiloxy ethyl methacrylate, trimethylsiloxy-ethylmethacrylate, acmc-1buxw, trimethylsilyloxyethyl-methacrylate, 2-trimethylsiloxyethyl methacrylate, 2-trimethylsiloxy ethylmethacrylate | |
| WUGOQZFPNUYUOO-UHFFFAOYSA-N | |
| 2-trimethylsilyloxyethyl 2-methylprop-2-enoate | |
| 87094 | |
| 94% |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 241-432-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only