missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)quinoline, 97%, Thermo Scientific™

Chemical Identifiers

CAS 347-42-2
Molecular Formula C10H6F3N
Molecular Weight (g/mol) 197.16
MDL Number MFCD01862002
InChI Key YZSRICFIQLVSMQ-UHFFFAOYSA-N
Synonym 2-trifluoromethyl quinoline, 2-trifluoromethyl-quinoline, pubchem23495, trifluoromethyl quinoline, quinoline,2-trifluoromethyl
PubChem CID 2777805
IUPAC Name 2-(trifluoromethyl)quinoline
SMILES FC(F)(F)C1=NC2=CC=CC=C2C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC432620010
SDS View Documents
Thermo Scientific Chemicals
432620010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

347-42-2
197.16
YZSRICFIQLVSMQ-UHFFFAOYSA-N
2777805
FC(F)(F)C1=NC2=CC=CC=C2C=C1
C10H6F3N
MFCD01862002
2-trifluoromethyl quinoline, 2-trifluoromethyl-quinoline, pubchem23495, trifluoromethyl quinoline, quinoline,2-trifluoromethyl
2-(trifluoromethyl)quinoline

Specifications

347-42-2
100.0
Authentic
Glass bottle
MFCD01862002
2-trifluoromethyl quinoline, 2-trifluoromethyl-quinoline, pubchem23495, trifluoromethyl quinoline, quinoline,2-trifluoromethyl
FC(F)(F)C1=NC2=CC=CC=C2C=C1
197.16
197.16
2-(Trifluoromethyl)quinoline
96.0
233.0°C
96% min. (GC)
C10H6F3N
1g
YZSRICFIQLVSMQ-UHFFFAOYSA-N
2-(trifluoromethyl)quinoline
2777805
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only