missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trifluoromethyl)quinoline, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 432620010
Specifications
| 2-(Trifluoromethyl)quinoline | |
| 96.0 | |
| 233.0°C | |
| 96% min. (GC) | |
| C10H6F3N | |
| 1g | |
| YZSRICFIQLVSMQ-UHFFFAOYSA-N | |
| 2-(trifluoromethyl)quinoline | |
| 2777805 | |
| 97% |
| 347-42-2 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| MFCD01862002 | |
| 2-trifluoromethyl quinoline, 2-trifluoromethyl-quinoline, pubchem23495, trifluoromethyl quinoline, quinoline,2-trifluoromethyl | |
| FC(F)(F)C1=NC2=CC=CC=C2C=C1 | |
| 197.16 | |
| 197.16 |
Chemical Identifiers
| 347-42-2 | |
| 197.16 | |
| YZSRICFIQLVSMQ-UHFFFAOYSA-N | |
| 2777805 | |
| FC(F)(F)C1=NC2=CC=CC=C2C=C1 |
| C10H6F3N | |
| MFCD01862002 | |
| 2-trifluoromethyl quinoline, 2-trifluoromethyl-quinoline, pubchem23495, trifluoromethyl quinoline, quinoline,2-trifluoromethyl | |
| 2-(trifluoromethyl)quinoline |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning
RUO â Research Use Only