missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-(Trifluoromethyl)pyrrolidine, 95%

Catalog No. p-7028506
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
Catalog No. Quantity
AAH25852MD 250 mg
AAH2585203 1 g
2 options

Catalog No. AAH25852MD

Supplier: Thermo Scientific Chemicals H25852MD

Only null left

CAS: 109074-67-1 | C5H8F3N | 139.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109074-67-1
Molecular Formula C5H8F3N
Molecular Weight (g/mol) 139.121
MDL Number MFCD02663405
InChI Key VINAMCOZNJHNIH-UHFFFAOYSA-N
Synonym 2-trifluoromethyl pyrrolidine, 2-trifluoromethyl-pyrrolidine, dl-2-trifluoromethylpyrrolidine, pyrrolidine,2-trifluoromethyl, pyrrolidine, 2-trifluoromethyl, +/--2-trifluoromethyl pyrrolidine, acmc-20dptq, acmc-1cih7, 2-trifluoromethyl tetrahydropyrrole
PubChem CID 2782839
IUPAC Name 2-(trifluoromethyl)pyrrolidine
SMILES C1CC(NC1)C(F)(F)F

Specifications

Density 1.21
Boiling Point 110°C to 112°C
Flash Point 26°C (78°F)
Refractive Index 1.379
Quantity 250 mg
UN Number UN2733
Sensitivity Air Sensitive
Formula Weight 139.12
Percent Purity 95%
Chemical Name or Material (+/-)-2-(Trifluoromethyl)pyrrolidine
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only