missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 368-48-9 |
|---|---|
| Molecular Formula | C6H4F3N |
| Molecular Weight (g/mol) | 147.10 |
| MDL Number | MFCD02183562 |
| InChI Key | ATRQECRSCHYSNP-UHFFFAOYSA-N |
| Synonym | 2-trifluoromethyl pyridine, 2-trifluoromethyl-pyridine, trifluoromethylpyridine, 2-1,1,1-trifluoromethyl pyridine, pyridine, trifluoromethyl, alpha,alpha,alpha-trifluoro-2-picoline, 2-cf3-pyridine, trifluoromethyl pyridine |
| PubChem CID | 136199 |
| IUPAC Name | 2-(trifluoromethyl)pyridine |
| SMILES | FC(F)(F)C1=CC=CC=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC442900010
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| C6H4F3N | |
| MFCD02183562 | |
| 2-trifluoromethyl pyridine, 2-trifluoromethyl-pyridine, trifluoromethylpyridine, 2-1,1,1-trifluoromethyl pyridine, pyridine, trifluoromethyl, alpha,alpha,alpha-trifluoro-2-picoline, 2-cf3-pyridine, trifluoromethyl pyridine | |
| 2-(trifluoromethyl)pyridine |
Specifications
| 368-48-9 | |
| 42°C | |
| C6H4F3N | |
| MFCD02183562 | |
| ATRQECRSCHYSNP-UHFFFAOYSA-N | |
| 2-(trifluoromethyl)pyridine | |
| 136199 | |
| 97% |
| 138°C to 141°C | |
| 96% min. (GC) | |
| 1.4183 | |
| 2-trifluoromethyl pyridine, 2-trifluoromethyl-pyridine, trifluoromethylpyridine, 2-1,1,1-trifluoromethyl pyridine, pyridine, trifluoromethyl, alpha,alpha,alpha-trifluoro-2-picoline, 2-cf3-pyridine, trifluoromethyl pyridine | |
| FC(F)(F)C1=CC=CC=N1 | |
| 147.10 | |
| 147.1 | |
| 2-(Trifluoromethyl)pyridine |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Warning
RUO – Research Use Only