missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenylhydrazine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7137404
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27665G 5 g
T276625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27665G Supplier TCI America Supplier No. T27665G

Chemical Identifiers

CAS 3107-34-4
Molecular Formula C7H8ClF3N2
Molecular Weight (g/mol) 212.6
MDL Number MFCD00102619
InChI Key ZUSWDTWYONAOPH-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenylhydrazine hydrochloride, 2-trifluoromethyl phenylhydrazine hcl, 2-hydrazinobenzotrifluoride hcl, 2-hydrazinobenzotrifluoride hydrochloride, 2-trifluoromethyl phenyl hydrazine hydrochloride, 2-trifluoromethyl phenylhydrazine monohydrochloride, 1-2-trifluoromethyl phenyl hydrazine hydrochloride, n'-2-trifluoromethyl-phenyl-hydrazinium, chloride, 2-trifluoromethylphenylhydrazine hydrochloride
PubChem CID 12891828
IUPAC Name [2-(trifluoromethyl)phenyl]hydrazine;hydrochloride
SMILES C1=CC=C(C(=C1)C(F)(F)F)NN.Cl

Specifications

Melting Point 223°C
Color White-Yellow
Quantity 5 g
Formula Weight 212.60
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)phenylhydrazine Hydrochloride