missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenylhydrazine hydrochloride, 98%

Catalog No. p-7028060
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
AAB2518206 5 g
AAB2518214 25 g
2 options

Catalog No. AAB2518206

Supplier: Thermo Scientific Chemicals B2518206

Only null left

CAS: 3107-34-4 | C7H8ClF3N2 | 212.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3107-34-4
Molecular Formula C7H8ClF3N2
Molecular Weight (g/mol) 212.6
MDL Number MFCD00102619
InChI Key ZUSWDTWYONAOPH-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenylhydrazine hydrochloride, 2-trifluoromethyl phenylhydrazine hcl, 2-hydrazinobenzotrifluoride hcl, 2-hydrazinobenzotrifluoride hydrochloride, 2-trifluoromethyl phenyl hydrazine hydrochloride, 2-trifluoromethyl phenylhydrazine monohydrochloride, 1-2-trifluoromethyl phenyl hydrazine hydrochloride, n'-2-trifluoromethyl-phenyl-hydrazinium, chloride, 2-trifluoromethylphenylhydrazine hydrochloride
PubChem CID 12891828
IUPAC Name [2-(trifluoromethyl)phenyl]hydrazine;hydrochloride
SMILES C1=CC=C(C(=C1)C(F)(F)F)NN.Cl

Specifications

Melting Point ∼235°C (decomposition)
Quantity 5 g
Beilstein 4582303
Sensitivity Hygroscopic
Formula Weight 212.6
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)phenylhydrazine hydrochloride
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only