missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenylhydrazine, 97%

Catalog No. p-7022150
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAA1478103 1 g
AAA1478106 5 g
2 options

Catalog No. AAA1478103

Supplier: Thermo Scientific Chemicals A1478103

Only null left

CAS: 365-34-4 | C7H7F3N2 | 176.142 g/mol

2-(Trifluoromethyl)phenylhydrazine is used as an intermediate in organic syntheses. It is also ued as a fine chemical intermediate, GC derivatizing agent, coupling reagent and N- protecting agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)phenylhydrazine is used as an intermediate in organic syntheses. It is also ued as a fine chemical intermediate, GC derivatizing agent, coupling reagent and N- protecting agent.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 365-34-4
Molecular Formula C7H7F3N2
Molecular Weight (g/mol) 176.142
MDL Number MFCD00052687
InChI Key JSWQDLBFVSTSIW-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenylhydrazine, 2-trifluoromethyl phenyl hydrazine, 2-trifluoromethylphenylhydrazine, 1-2-trifluoromethyl phenyl hydrazine, m-trifluoromethylphenylhydrazine, hydrazine, 2-trifluoromethyl phenyl, buttpark 44\57-17, 2-trifluoromethyl-phenyl-hydrazine
PubChem CID 520800
IUPAC Name [2-(trifluoromethyl)phenyl]hydrazine
SMILES C1=CC=C(C(=C1)C(F)(F)F)NN

Specifications

Melting Point 57°C to 63°C
Quantity 1 g
Beilstein 641592
Solubility Information Insoluble in water.
Formula Weight 176.14
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)phenylhydrazine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only