missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenylglyoxal hydrate, 98%, dry wt. basis

Catalog No. aal20280md
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL20280MD 250 mg
AAL2028003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL20280MD Supplier Thermo Scientific Chemicals Supplier No. L20280MD
Only null left

CAS: 745783-91-9 | C9H5F3O2 | 202.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 745783-91-9
Molecular Formula C9H5F3O2
Molecular Weight (g/mol) 202.13
MDL Number MFCD08533288
InChI Key HBOSPPHIPGYHDI-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenylglyoxal hydrate, 2-oxo-2-2-trifluoromethyl phenyl acetaldehyde hydrate, 2-oxo-2-2-trifluoromethyl phenyl ethanal, oxamethane, benzeneacetaldehyde, a-oxo-2-trifluoromethyl-,hydrate 1:1
PubChem CID 2783281
IUPAC Name 2-oxo-2-[2-(trifluoromethyl)phenyl]acetaldehyde;hydrate
SMILES FC(F)(F)C1=CC=CC=C1C(=O)C=O

Specifications

Melting Point 36°C to 43°C
Quantity 250 mg
Formula Weight 202.13 (Anhydrous)
Percent Purity 98%
Assay (dry wt.)
Chemical Name or Material 2-(Trifluoromethyl)phenylglyoxal hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only