missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenylacetonitrile 98.0+%, TCI America™

Catalog No. T23665G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23665G 5 g
T236625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23665G Supplier TCI America Supplier No. T23665G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3038-47-9
Molecular Formula C9H6F3N
Molecular Weight (g/mol) 185.15
MDL Number MFCD00001903
InChI Key QXDCZSJGEUSERL-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenylacetonitrile, 2-2-trifluoromethyl phenyl acetonitrile, 2-trifluoromethyl benzyl cyanide, o-trifluoromethylbenzyl cyanide, 2-trifluoromethylbenzyl cyanide, benzeneacetonitrile, 2-trifluoromethyl, o-trifluoromethylphenylacetonitrile, 2-trifluoromethyl phenyl acetonitrile
PubChem CID 76435
IUPAC Name 2-[2-(trifluoromethyl)phenyl]acetonitrile
SMILES FC(F)(F)C1=CC=CC=C1CC#N

Specifications

Color White-Yellow
Boiling Point 92°C
Quantity 5 g
UN Number 3439
Formula Weight 185.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)phenylacetonitrile