Learn More
2-(Trifluoromethyl)phenyl isothiocyanate, 98%
CAS: 1743-86-8 | C8H4F3NS | 203.182 g/mol
$54.56 - $183.59
Chemical Identifiers
| CAS | 1743-86-8 |
|---|---|
| Molecular Formula | C8H4F3NS |
| Molecular Weight (g/mol) | 203.182 |
| MDL Number | MFCD00039644 |
| InChI Key | FCEKLQPJGXIQRY-UHFFFAOYSA-N |
| Synonym | 2-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl benzene, 2-trifluoromethylphenylisothiocyanate, 2-trifluoromethyl phenylisothiocyanate, 2-trifluoromethylphenyl isothiocyanate, o-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl-benzene, benzene, 1-isothiocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isothiocyanate, isothiocyanic acid 2-trifluoromethyl phenyl ester |
| PubChem CID | 737162 |
| IUPAC Name | 1-isothiocyanato-2-(trifluoromethyl)benzene |
| SMILES | C1=CC=C(C(=C1)C(F)(F)F)N=C=S |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1058603
|
Thermo Scientific Chemicals
L1058603 |
1 g |
Each for $54.56
|
|
|||||
|
AAL1058606
|
Thermo Scientific Chemicals
L1058606 |
5 g |
Each for $183.59
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1743-86-8 | |
| 203.182 | |
| FCEKLQPJGXIQRY-UHFFFAOYSA-N | |
| 737162 | |
| C1=CC=C(C(=C1)C(F)(F)F)N=C=S |
| C8H4F3NS | |
| MFCD00039644 | |
| 2-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl benzene, 2-trifluoromethylphenylisothiocyanate, 2-trifluoromethyl phenylisothiocyanate, 2-trifluoromethylphenyl isothiocyanate, o-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl-benzene, benzene, 1-isothiocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isothiocyanate, isothiocyanic acid 2-trifluoromethyl phenyl ester | |
| 1-isothiocyanato-2-(trifluoromethyl)benzene |
Specifications
| 1743-86-8 | |
| 216°C | |
| Pungent | |
| 1.566 | |
| 1 g | |
| 2693552 | |
| 2-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl benzene, 2-trifluoromethylphenylisothiocyanate, 2-trifluoromethyl phenylisothiocyanate, 2-trifluoromethylphenyl isothiocyanate, o-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl-benzene, benzene, 1-isothiocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isothiocyanate, isothiocyanic acid 2-trifluoromethyl phenyl ester | |
| C1=CC=C(C(=C1)C(F)(F)F)N=C=S | |
| 203.182 | |
| 203.19 | |
| 2-(Trifluoromethyl)phenyl isothiocyanate |
| 1.346 | |
| 105°C (221°F) | |
| C8H4F3NS | |
| MFCD00039644 | |
| UN2922 | |
| Moisture sensitive | |
| FCEKLQPJGXIQRY-UHFFFAOYSA-N | |
| 1-isothiocyanato-2-(trifluoromethyl)benzene | |
| 737162 | |
| 98% |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only