missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenyl isothiocyanate, 98%

Catalog No. AAL1058603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1058603 1 g
AAL1058606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1058603 Supplier Thermo Scientific Chemicals Supplier No. L1058603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1743-86-8 | C8H4F3NS | 203.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1743-86-8
Molecular Formula C8H4F3NS
Molecular Weight (g/mol) 203.182
MDL Number MFCD00039644
InChI Key FCEKLQPJGXIQRY-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl benzene, 2-trifluoromethylphenylisothiocyanate, 2-trifluoromethyl phenylisothiocyanate, 2-trifluoromethylphenyl isothiocyanate, o-trifluoromethyl phenyl isothiocyanate, 1-isothiocyanato-2-trifluoromethyl-benzene, benzene, 1-isothiocyanato-2-trifluoromethyl, alpha,alpha,alpha-trifluoro-o-tolyl isothiocyanate, isothiocyanic acid 2-trifluoromethyl phenyl ester
PubChem CID 737162
IUPAC Name 1-isothiocyanato-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)N=C=S

Specifications

Density 1.346
Boiling Point 216°C
Flash Point 105°C (221°F)
Odor Pungent
Refractive Index 1.566
Quantity 1 g
UN Number UN2922
Beilstein 2693552
Sensitivity Moisture sensitive
Formula Weight 203.19
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)phenyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only