missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenol, 98%

CAS: 444-30-4 | C7H5F3O | 162.111 g/mol

Supplier:  Thermo Scientific Chemicals A1423103

 View more versions of this product

Catalog No. AAA1423103

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-(Trifluoromethyl)phenol
45°C to 46°C
65°C (149°F)
C7H5F3O
1 g
1867917
ZOQOPXVJANRGJZ-UHFFFAOYSA-N
2-(trifluoromethyl)phenol
67958
98%
444-30-4
147°C to 148°C
Phenol-like
MFCD00002222
UN3261
2-trifluoromethyl phenol, 2-hydroxybenzotrifluoride, o-trifluoromethylphenol, o-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-o-cresol, phenol, 2-trifluoromethyl, o-trifluoromethoxy phenol, a,a,a-trifluoro-o-cresol, 2-hydroxy-alpha,alpha,alpha-trifluorotoluene
C1=CC=C(C(=C1)C(F)(F)F)O
162.111
162.11

Chemical Identifiers

444-30-4
162.111
ZOQOPXVJANRGJZ-UHFFFAOYSA-N
67958
C1=CC=C(C(=C1)C(F)(F)F)O
C7H5F3O
MFCD00002222
2-trifluoromethyl phenol, 2-hydroxybenzotrifluoride, o-trifluoromethylphenol, o-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-o-cresol, phenol, 2-trifluoromethyl, o-trifluoromethoxy phenol, a,a,a-trifluoro-o-cresol, 2-hydroxy-alpha,alpha,alpha-trifluorotoluene
2-(trifluoromethyl)phenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c

H302+H312+H332-H318-H335

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 207-148-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only