missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenol, 98%

Catalog No. AAA1423114
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1423114 25 g
AAA1423103 1 g
AAA1423106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1423114 Supplier Thermo Scientific Chemicals Supplier No. A1423114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 444-30-4 | C7H5F3O | 162.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 444-30-4
Molecular Formula C7H5F3O
Molecular Weight (g/mol) 162.111
MDL Number MFCD00002222
InChI Key ZOQOPXVJANRGJZ-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenol, 2-hydroxybenzotrifluoride, o-trifluoromethylphenol, o-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-o-cresol, phenol, 2-trifluoromethyl, o-trifluoromethoxy phenol, a,a,a-trifluoro-o-cresol, 2-hydroxy-alpha,alpha,alpha-trifluorotoluene
PubChem CID 67958
IUPAC Name 2-(trifluoromethyl)phenol
SMILES C1=CC=C(C(=C1)C(F)(F)F)O

Specifications

Melting Point 45°C to 46°C
Boiling Point 147°C to 148°C
Flash Point 65°C (149°F)
Odor Phenol-like
Quantity 25 g
UN Number UN3261
Beilstein 1867917
Formula Weight 162.11
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)phenol
Hazard Category H302+H312+H332-H318-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P363-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 207-148-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only