missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzylamine, 98%

Catalog No. AC312950010
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC312950010 1 g Glass bottle
AC312950050 5 g Glass bottle
AC312950100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC312950010 Supplier Thermo Scientific Chemicals Supplier No. 312950010

CAS: 3048-01-9 | C8H8F3N | 175.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3048-01-9
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.16
MDL Number MFCD00010297
InChI Key ZSKQIFWUTUZAGF-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzylamine, 2-trifluoromethylbenzylamine, 2-trifluoromethyl phenyl methanamine, o-trifluoromethylbenzyl amine, o-trifluoromethylbenzylamine, 2-trifluoromethyl-benzylamine, 1-2-trifluoromethyl phenyl methanamine, benzenemethanamine, 2-trifluoromethyl, o-trifluoromethyl benzylamine
PubChem CID 76447
IUPAC Name [2-(trifluoromethyl)phenyl]methanamine
SMILES C1=CC=C(C(=C1)CN)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2490g/mL
Flash Point 69°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CF3C6H4CH2NH2
Refractive Index 1.4700 to 1.472
Quantity 1 g
Specific Gravity 1.249
Formula Weight 175.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethyl)benzylamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 221-258-2
TSCA TSCA

RUO – Research Use Only