missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzoyl Chloride 98.0+%, TCI America™

Catalog No. T128025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T128025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T128025G Supplier TCI America Supplier No. T128025G

Chemical Identifiers

CAS 312-94-7
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.56
MDL Number MFCD00000667
InChI Key MXIUWSYTQJLIKE-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzoyl chloride, o-trifluoromethylbenzoyl chloride, alpha,alpha,alpha-trifluoro-o-toluoyl chloride, 2-trifluoromethyl benzoylchloride, unii-d01d0b64eq, otf-boc, 2-trifluoromethyl-benzoyl chloride, o-trifluoromethyl benzoyl chloride
PubChem CID 67561
IUPAC Name 2-(trifluoromethyl)benzoyl chloride
SMILES FC(F)(F)C1=CC=CC=C1C(Cl)=O

Specifications

Color Yellow
Boiling Point 90°C
Quantity 25 g
UN Number 3265
Formula Weight 208.56
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethyl)benzoyl Chloride
EINECSNumber (3)-3798
TSCA No