missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzonitrile 98.0+%, TCI America™

Catalog No. C10045G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10045G 5 g
C100425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10045G Supplier TCI America Supplier No. C10045G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 447-60-9
Molecular Formula C8H4F3N
Molecular Weight (g/mol) 171.122
MDL Number MFCD00001791
InChI Key SOZGHDCEWOLLHV-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzonitrile, o-trifluoromethylbenzonitrile, 2-cyanobenzotrifluoride, alpha,alpha,alpha-trifluoro-o-tolunitrile, a,a,a-trifluoro-o-tolunitrile, 2-trifluoromethyl-benzonitrile, alpha,alpha,alpha-trifluoro-2-toluonitrile, benzonitrile, trifluoromethyl, benzonitrile, 2-trifluoromethyl, 2-trifluoromethyl benzenecarbonitrile
PubChem CID 67972
IUPAC Name 2-(trifluoromethyl)benzonitrile
SMILES C1=CC=C(C(=C1)C#N)C(F)(F)F

Specifications

Color Yellow
Boiling Point 206°C
Quantity 5 g
UN Number 3276
Formula Weight 171.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethyl)benzonitrile