missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzonitrile, 97%

Catalog No. AAA1495106
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1495106 5 g
AAA1495109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1495106 Supplier Thermo Scientific Chemicals Supplier No. A1495106

CAS: 447-60-9 | C8H4F3N | 171.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 447-60-9
Molecular Formula C8H4F3N
Molecular Weight (g/mol) 171.122
MDL Number MFCD00001791
InChI Key SOZGHDCEWOLLHV-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzonitrile, o-trifluoromethylbenzonitrile, 2-cyanobenzotrifluoride, alpha,alpha,alpha-trifluoro-o-tolunitrile, a,a,a-trifluoro-o-tolunitrile, 2-trifluoromethyl-benzonitrile, alpha,alpha,alpha-trifluoro-2-toluonitrile, benzonitrile, trifluoromethyl, benzonitrile, 2-trifluoromethyl, 2-trifluoromethyl benzenecarbonitrile
PubChem CID 67972
IUPAC Name 2-(trifluoromethyl)benzonitrile
SMILES C1=CC=C(C(=C1)C#N)C(F)(F)F

Specifications

Melting Point 17°C to 18°C
Density 1.294
Boiling Point 204°C to 206°C
Flash Point 90°C (194°F)
Refractive Index 1.4632
Quantity 5 g
UN Number UN3276
Beilstein 2048152
Formula Weight 171.12
Percent Purity 97%
Chemical Name or Material 2-(Trifluoromethyl)benzonitrile
Show More Show Less
Hazard Category H227-H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 207-184-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only