missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzoic Acid 98.0+%, TCI America™

Catalog No. T130210G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T130210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T130210G Supplier TCI America Supplier No. T130210G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 433-97-6
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00002476
InChI Key FBRJYBGLCHWYOE-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzoic acid, o-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-o-toluic acid, o-carboxybenzotrifluoride, 2-carboxybenzotrifluoride, unii-0r1ck7mzua, trifluoromethylbenzoic acid, 2-trifluoromethyl benzoate, 2-trifluromethylbenzoic acid, trifluoromethyl benzoic acid
PubChem CID 9899
ChEBI CHEBI:60694
IUPAC Name 2-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC=CC=C1C(F)(F)F

Specifications

Melting Point 110°C
Boiling Point 247°C
Quantity 10 g
Formula Weight 190.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)benzoic Acid
EINECSNumber (3)-1414
TSCA No