missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog No. AC188450050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC188450050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC188450050 Supplier Thermo Scientific Chemicals Supplier No. 188450050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 433-97-6 | Molecular Formula: C8H5F3O2 | Molecular Weight: 190.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 433-97-6
Molecular Formula C8H5F3O2
Molecular Weight (g/mol) 190.12
MDL Number MFCD00002476
InChI Key FBRJYBGLCHWYOE-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzoic acid, o-trifluoromethylbenzoic acid, alpha,alpha,alpha-trifluoro-o-toluic acid, o-carboxybenzotrifluoride, 2-carboxybenzotrifluoride, unii-0r1ck7mzua, trifluoromethylbenzoic acid, 2-trifluoromethyl benzoate, 2-trifluromethylbenzoic acid, trifluoromethyl benzoic acid
PubChem CID 9899
ChEBI CHEBI:60694
IUPAC Name 2-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC=CC=C1C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 107.0°C to 110.0°C
Color Brown-Yellow to Yellow
Boiling Point 247.0°C (753.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CF3C6H4CO2H
Quantity 5 g
Beilstein 09, III, 2308
Formula Weight 190.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)benzoic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 207-093-9
TSCA TSCA

RUO – Research Use Only